Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD47 Protein, Fc (L234A, L235A, P329G),Avitag™
Manufacturer:  ACROBiosystems CD7H82F825UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD47, Fc (L234A, L235A, P329G),Avitag (CD7-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # Q08722-3).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.9 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Mouse IgG1 Fc Tag (HPLC-verified) (Cat. No. SIA-H52A8) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD47, Fc (L234A, L235A, P329G),Avitag (Cat. No. CD7-H82F8) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human CD47 Antibody, Human IgG4 at 2 μg/mL (100 μL/well) can bind Biotinylated Human CD47, Fc (L234A, L235A, P329G),Avitag (Cat. No. CD7-H82F8) with a linear range of 0.2-5 ng/mL (Routinely tested).
Catalog No. 50-195-6682