Learn More
ACROBiosystems ACROBiosystems Human TRAIL / TNFRSF10 Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems TRLH5259100UG
Recombinant Protein;100UG;Human TRAIL, Mouse IgG2a Fc Tag (TRL-H5259) is expressed from human 293 cells (HEK293). It contains AA Val 114 - Gly 281 (Accession # P50591-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 46.4 kDa. The protein migrates as 47-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Osteoprotegerin, His Tag (Cat. No. TNB-H5220) at 5 μg/mL (100 μL/well) can bind Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) with a linear range of 0.8-13 ng/mL (QC tested).;Immobilized Human TRAIL R1, His Tag (Cat. No. TR1-H5228) at 5 μg/mL (100 μL/well) can bind Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) with a linear range of 2-20 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.