Learn More
ACROBiosystems ACROBiosystems Biotinylated Human VSIG3 Protein, Fc,Avitag™
Supplier:  ACROBiosystems VS3H82F925UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human VSIG3, Fc,Avitag (VS3-H82F9) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 241 (Accession # Q5DX21-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.5 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human VSIG3, Fc,Avitag (Cat. No. VS3-H82F9) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human B7-H5, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. B75-H5258) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-6837