Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Biotinylated Human PVRIG Protein, Fc,Avitag™  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PVGH82F625UG
Recombinant Protein;25UG;Biotinylated Human PVRIG, Fc,Avitag (PVG-H82F6) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 41.9 kDa. The protein migrates as 46-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) at 10 μg/mL (100 μL/well) can bind Biotinylated Human PVRIG, Fc,Avitag (Cat. No. PVG-H82F6) with a linear range of 5-156 ng/mL (QC tested).;Immobilized Human Nectin-2, His Tag (Cat. No. PV2-H52E2) at 10 μg/mL (100 μL/well) can bind Biotinylated Human PVRIG, Fc,Avitag (Cat. No. PVG-H82F6) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-195-6852