Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Human IL-5 R alpha / CD125 Protein, Fc Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ILAH525950UG
Recombinant Protein;50UG;Human IL-5 R alpha, Fc Tag (ILA-H5259) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Trp 342 (Accession # Q01344-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.2 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-5, His Tag (Cat. No. IL5-H52H3) at 5 μg/mL (100 μL/well) can bind Human IL-5 R alpha, Fc Tag (Cat. No. ILA-H5259) with a linear range of 2-39 ng/mL (QC tested).
Catalog No. 50-195-6867