Learn More
ACROBiosystems ACROBiosystems Mouse IL-2 R alpha / CD25 Protein, His Tag
Supplier:  ACROBiosystems ILAM52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse IL-2 R alpha, His Tag (ILA-M52H9) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Lys 236 (Accession # P01590-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-2, Fc Tag, Avi Tag (Cat. No. IL2-H82F3) at 5 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Mouse IL-2 R alpha, His Tag (Cat. No. ILA-M52H9) with a linear range of 5-40 ng/mL (QC tested).;Immobilized Mouse IL-2 R alpha, His Tag (Cat. No. ILA-M52H9) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-2, Fc Tag, Avi Tag (Cat. No. IL2-H82F3) with a linear range of 0.6-10 ng/mL (Routinely tested).
Catalog No. 50-195-6880