Learn More
ACROBiosystems ACROBiosystems Cynomolgus Complement Component C5a Protein, Tag Free
Supplier: ACROBiosystems C5AC5118100UG
Recombinant Protein;100UG;Cynomolgus C5a, Tag Free (C5A-C5118) is expressed from E.coli cells. It contains AA Met 678 - Arg 751 (Accession # XP_015292262.1).;This protein carries no "tag". The protein has a calculated MW of 8.4 kDa. The protein migrates as 11 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus C5a, Tag Free (Cat. No. C5A-C5118) at 2 μg/mL (100 μL/well) can bind Anti-C5a (Human IgG1) with a linear range of 0.8-13 ng/mL (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.