Learn More
ACROBiosystems ACROBiosystems Mouse IgG1 Fc Protein, Tag Free
Supplier:  ACROBiosystems IG1M5208200UG
Encompass_Preferred
Recombinant Protein;200UG;Mouse IgG Fc, Tag Free (IG1-M5208) is expressed from human 293 cells (HEK293). It contains AA Val 111 - Lys 324 (Accession # AAK53870.1).;This protein carries no "tag". The protein has a calculated MW of 24.3 kDa. The protein migrates as 28-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6975