Learn More
ACROBiosystems ACROBiosystems Human Complement Factor D / CFD Protein, His Tag
Supplier: ACROBiosystems CFDH52H850UG
Recombinant Protein;50UG;Human Complement Factor D, His Tag (CFD-H52H8) is expressed from human 293 cells (HEK293) and active. It contains AA Ile 26 - Ala 253 (Accession # P00746-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 28 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.