Learn More
ACROBiosystems ACROBiosystems RFP Fusion Human Mesothelin / MSLN (296-580) Protein, His Tag
Supplier: ACROBiosystems MSNHR2P425UG
Recombinant Protein;25UG;RFP Fusion Human Mesothelin (296-580), His Tag (MSN-HR2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296-Gly 580 (Accession # AAH09272.1).;This protein carries a RFP tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 62.0 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human MSLN Mab, Human IgG1 at 10 μg/mL (100 μL/well) can bind RFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HR2P4) with a linear range of 0.078-0.625 μg/mL (QC tested).;can bind Human MS4A1 / CD20 Full Length Protein, His Tag (Cat. No. CD0-H55Ha)with an affinity constant of 1.21 nM as determined in a SPR assay (Biacore T200) (Routinely tested).;2e5 of the MSLN-CAR-293 cells were stained with 100 μL of 3 μg/mL of RFP Fusion Human Mesothelin (296-580), His Tag (Cat. No. MSN-HR2P4). Non-transfected 293 cells were used as negative control (QC tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.