Learn More
ACROBiosystems ACROBiosystems Human TrkA / NTRK1 (33-417) Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems TRAH5259100UG
Recombinant Protein;100UG;Human TrkA (33-417), Mouse IgG2a Fc Tag (TRA-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 33 - Gly 417 (Accession # P04629-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.2 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.