Learn More
ACROBiosystems ACROBiosystems Biotinylated Human B7-H3 (4Ig) / B7-H3b Protein, His,Avitag™
Supplier: ACROBiosystems B7BH82E825UG
Recombinant Protein;25UG;Biotinylated Human B7-H3 (4Ig), His,Avitag (B7B-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.3 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human B7-H3 (4Ig), His,Avitag (Cat. No. B7B-H82E8) with a linear range of 0.3-2 ng/mL (QC tested).;Immobilized Biotinylated Human B7-H3 (4Ig), His,Avitag (Cat. No. B7B-H82E8) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human B7-H3 / B7-H3 (4Ig) Antibody, Human IgG1 with a linear range of 0.3-5 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.