Learn More
ACROBiosystems ACROBiosystems Human TIGIT Protein, Fc Tag
Supplier:  ACROBiosystems TITH5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Human TIGIT, Fc Tag (TIT-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession # Q495A1 -1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 39.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD155, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD5-H5254) at 5 μg/mL (100 μL/well) can bind Human TIGIT, Fc Tag (Cat. No. TIT-H5254) with a linear range of 0.6-5 ng/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.313-10 μg/mL (Routinely tested).;FACS assay shows that recombinant Human TIGIT, Fc Tag (Cat. No. TIT-H5254) can bind to 293T cell overexpressing human CD155. The concentration of TIGIT used is 0.1 μg/mL (Routinely tested).
Catalog No. 50-195-7120