Learn More
ACROBiosystems ACROBiosystems Rhesus macaque OX40 / TNFRSF4 / CD134 Protein, His Tag
Supplier:  ACROBiosystems OX0C5220100UG
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque OX40, His Tag (OX0-C5220) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Ala 214 (Accession # XP_001090870.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.1 kDa. The protein migrates as 33-50 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Rhesus macaque OX40, His Tag (Cat. No. OX0-C5220) at 2 μg/mL (100 μl/well) can bind Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) with a linear range of 0.2-25 ng/mL (QC tested).
Catalog No. 50-195-7241