Learn More
ACROBiosystems ACROBiosystems Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] Hemagglutinin 1 (HA1) Protein, His Tag,Avitag™
Manufacturer:  ACROBiosystems HA1V82E225UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] HA1, His Tag,Avitag (HA1-V82E2) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.3 kDa. The protein migrates as 47-70 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7246