Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Rat GITR / TNFRSF18 Protein, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner GIRR5222100UG
Encompass_Preferred
Recombinant Protein;100UG;Rat GITR, His Tag (GIR-R5222) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Lys 121 (Accession # Q5M835).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 12.3 kDa. The protein migrates as 21 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7285