Learn More
ACROBiosystems ACROBiosystems Mouse IL-23 R Protein, Fc Tag
Supplier: ACROBiosystems ILRM5251100UG
Recombinant Protein;100UG;Mouse IL-23R, Fc Tag (ILR-M5251) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asp 372 (Accession # NP_653131).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 66.8 kDa. As a result of glycosylation, the protein migrates as 90-100 kDa under reducing (R) condition, and 180-200 kDa under non-reducing (NR) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.