Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] Hemagglutinin (HA) Protein, Avitag™,His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner HA1V82E125UG
Recombinant Protein;25UG;Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] HA, Avitag,His Tag (HA1-V82E1) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Lys 523 (Accession # AAO46082.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 60.0 kDa. The protein migrates as 66-70 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Anti-Influenza HA MAb CM2S3 at 2 μg/mL (100 μL/well) can bind Biotinylated Influenza A [A/Guinea fowl/Hong Kong/WF10/99(H9N2)] HA, Avitag,His Tag (Cat. No. HA1-V82E1) with a linear range of 2-63 ng/mL (QC tested).
Catalog No. 50-195-7356