Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Mouse B7-H3 / CD276 Protein, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner B73M52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse B7-H3, His Tag (B73-M52H4) is expressed from human 293 cells (HEK293). It contains AA Val 29 - Phe 244 (Accession # NP_598744).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7374