Learn More
ACROBiosystems ACROBiosystems Biotinylated Human 4-1BB /TNFRSF9 Protein, Fc,Avitag™
Manufacturer:  ACROBiosystems 41BH82F725UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human 4-1BB, Fc,Avitag (41B-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Gln 186 (Accession # NP_001552.2).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 45.7 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4‑1BB Ligand, Fc Tag (Cat. No. 41L-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human 4-1BB, Fc,Avitag (Cat. No. 41B-H82F7) with a linear range of 2-19 ng/mL (QC tested).;Flow Cytometry assay shows that Biotinylated Human 4-1BB, Fc,Avitag (Cat. No. 41B-H82F7) can bind to 293T cells overexpressing Human 4-1BBL. The concentration of 4-1BB used is 0.3 μg/mL (Routinely tested).
Catalog No. 50-195-7376