Learn More
ACROBiosystemsSupplier Diversity Partner ACROBiosystems Biotinylated Human BTLA (31-134) Protein, Fc,Avitag™  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner BTAH82F825UG
Recombinant Protein;25UG;Biotinylated Human BTLA (31-134), Fc,Avitag (BTA-H82F8) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # Q7Z6A9).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 40.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 10 μg/mL (100 μL/well) can bind Biotinylated Human BTLA (31-134), Fc,Avitag (Cat. No. BTA-H82F8) with a linear range of 0.3-5 μg/mL (QC tested).
Catalog No. 50-195-7406