Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-17 RA / CD217 Protein, Fc,Avitag™
Supplier:  ACROBiosystems ILAH82F125UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-17 RA, Fc,Avitag (ILA-H82F1) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 61.9 kDa. As a result of glycosylation, the protein migrates as 80-115 kDa under reducing (R) condition, and 150-200 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human IL17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17 RA, Fc,Avitag (Cat. No. ILA-H82F1) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-7423