Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Mesothelin / MSLN (296-580) Protein, His Tag, ultra sensitivity (primary amine labeling)
Supplier: ACROBiosystems MSNH822325UG
Recombinant Protein;25UG;Biotinylated Human Mesothelin (296-580), His Tag, primary amine labeling (MSN-H8223) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession # AAH09272). It is the biotinylated form of Human Mesothelin (296-580), His Tag (MSN-H5223).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33.0 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.