Learn More
ACROBiosystems ACROBiosystems ActiveMax Human VEGF165 Protein, Tag Free (MALS verified)
Supplier: ACROBiosystems VE5H421020UG
Recombinant Protein;20UG;ActiveMax Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries no "tag". The protein has a calculated MW of 19.2 kDa. As a result of glycosylation, the protein migrates as 24 kDa (monomer) under reducing (R) condition, and 43-50 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 0.01 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-31 ng/mL (QC tested).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R2, Avitag,His Tag (Cat. No. KDR-H82E5) with a linear range of 10-156 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.