Learn More
ACROBiosystems ACROBiosystems ActiveMax Human Oncostatin M (OSM) Protein, Tag Free
Supplier:  ACROBiosystems OSMH521350UG
Encompass_Preferred
Recombinant Protein;50UG;ActiveMax Human Oncostatin M, Tag Free (OSM-H5213) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Arg 252 (Accession # NP_065391.1).;This protein carries no "tag". The protein has a calculated MW of 25.8 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human Oncostatin M, Tag Free (Cat. No. OSM-H5213) at 5 μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 0.01-0.156 μg/mL (QC tested).
Catalog No. 50-195-7557