Learn More
ACROBiosystems ACROBiosystems ActiveMax Human LIF Protein, Tag Free
Supplier:  ACROBiosystems LIFH521B20UG
Encompass_Preferred
Recombinant Protein;20UG;ActiveMax Human LIF, Tag Free (LIF-H521b) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Phe 202 (Accession # AAH69540.1).;This protein carries no "tag". The protein has a calculated MW of 19.9 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human LIF, Tag Free (Cat. No. LIF-H521b) at 0.2 μg/mL (100 μL/well) can bind Human LIF R, Fc Tag (Cat. No. LIR-H4252) with a linear range of 0.016-0.25 μg/mL (QC tested).
Catalog No. 50-195-7558
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.