Learn More
ACROBiosystems ACROBiosystems ActiveMax Human Flt-3 Ligand Protein, Tag Free
Supplier:  ACROBiosystems FLLH521820UG
Encompass_Preferred
Recombinant Protein;20UG;ActiveMax Human Flt-3 Ligand, Tag Free (FLL-H5218) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Pro 185 (Accession # AAI26294).;This protein carries no "tag". The protein has a calculated MW of 18.0 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE).;Immobilized ActiveMax Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 1 μg/mL (100 μL/well) can bind Human Flt-3, Fc Tag (Cat. No. FL3-H5258) with a linear range of 2-31 ng/mL (QC tested).
Catalog No. 50-195-7566
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.