Learn More
ACROBiosystems ACROBiosystems ActiveMax Human Activin A / INHBA Protein, Tag Free, low endotoxin
Supplier:  ACROBiosystems ACAH421B25UG
Encompass_Preferred
Recombinant Protein;25UG;ActiveMax Human Activin A, Tag Free, low endotoxin (ACA-H421b) is expressed from human 293 cells (HEK293). It contains AA Gly 311 - Ser 426 (Accession # AAH07858.1).;This protein carries no "tag". The protein has a calculated MW of 13.0 kDa. the protein migrates as 14 kDa under reducing (R) condition, and 27 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human Activin A, Tag Free, low endotoxin (Cat. No. ACA-H421b) at 10 μg/mL (100 μL/well) can bind Human ACVR1B, Fc Tag (Cat. No. ACB-H5251) with a linear range of 0.313-5 μg/mL (Routinely tested).;The bio-activity of ActiveMax Human Activin A, Tag Free, low endotoxin (Cat. No. ACA-H421b) was determined by dose-dependent inhibition of the proliferation of MPC-11 cells. The EC50 for this effect is typically ≤ 9.5 ng/mL (Routinely tested).
Catalog No. 50-195-7569