Learn More
NEOGEN D/E Neutralizing Broth without Tween, 500 g
Supplier: NEOGEN 700003316
D/E Neutralizing Broth is used to neutralize and determine the bactericidal activity of antiseptics and disinfectants. D/E Neutralizing Broth without Tween is not intended for use in the diagnosis of disease or other conditions in humans.D/E Neutralizing Broth was developed by Dey and Engley to neutralize a broad spectrum of disinfectants and preservative antimicrobial chemicals, including quaternary ammonium compounds, phenolics, iodine, chlorine preparations, mercurials, formaldehyde, and glutaraldehyde. D/E Neutralizing media neutralize higher concentrations of residual antimicrobials when compared with other standard neutralizing formulations, such as Letheen media, Thioglycollate media, and Neutralizing Buffer.For more information visit https://www.neogen.com/solutions/Microbiology/de-neutralizing-broth-without-tween
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.