Learn More
ACROBiosystems FITC-Labeled Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control
Supplier:  ACROBiosystems DNPFM125UG
Encompass_Preferred
Antibody;25UG;Mouse IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. FITC-Labeled Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the FITC labeled form ofMonoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control (Cat. No. DNP-M1).; ;2e5 of Anti-CD19 CAR-293 cells were stained with a series of concentrations of FITC-Labeled Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-FM1), and FITC signal was used to evaluate the binding activity (QC tested). ;5e5 of PBMCs were stained with a series of concentrations of FITC-Labeled Anti-DNP Antibody, Mouse IgG1 (Cat. No. DNP-FM1), and FITC sign
Catalog No. 50-201-9410