Learn More
ACROBiosystemsSupplier Diversity Partner FITC-Labeled Monoclonal Anti-DNP antibody, Human IgG1 Isotype Control  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner DNPFM225UG
Antibody;25UG;Human IgG1/kappa;Specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.;Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies. FITC-Labeled Monoclonal Anti-DNP antibody, Mouse IgG1 Isotype Control was purified from HEK293 cell culture. It is the FITC labeled form ofMonoclonal Anti-DNP antibody, Human IgG1 Isotype Control (Cat. No. DNP-M2).;
Catalog No. 50-201-9412