Learn More
ACROBiosystemsSupplier Diversity Partner MERS Nucleocapsid protein, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner NUNM52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;MERS Nucleocapsid protein, His Tag (NUN-M52H5) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Asp 413 (Accession # K0BVN3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.7 kDa.
Catalog No. 50-201-9461