Learn More
ACROBiosystems Human Netrin receptor DCC / DCC Protein, His Tag
Supplier:  ACROBiosystems NECH52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Netrin receptor DCC, His Tag (NEC-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 119.5 kDa.
Catalog No. 50-201-9516