Learn More
ACROBiosystemsSupplier Diversity Partner Anti-SARS-CoV-2 Spike S1 Antibody, Chimeric mAb  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner S1NM130100UG
Antibody;100UG;Human IgG1/kappa;The cross-reactivity with other coronaviruses has not been tested yet.;Anti-SARS-CoV-2 Spike S1 Antibody, Chimeric MAb (S1N-M130) is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified recombinant SARS-CoV-2 Spike S1 Protein.
It is recommended to use antibody (S1N-M130) and (S1N-M122) as a pair set to detect SARS-CoV-2 Spike S1 in ELISA or serological assays.; ;Immobilized SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H2) at 2 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike S1 Antibody, chimeric mAb (Cat. No. S1N-M130) with a linear range of 0.08-2.5 ng/mL (QC tested).
Catalog No. 50-201-9524