Learn More
ACROBiosystems Human ROR1 Protein, Mouse IgG2a Fc Tag
Supplier: ACROBiosystems RO1H5256100UG
Recombinant Protein;100UG;Human ROR1, Mouse IgG2a Fc Tag (RO1-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.9 kDa.