Learn More
ACROBiosystemsSupplier Diversity Partner Human PCSK9 Protein, Mouse IgG2a Fc Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PC9H525750UG
Encompass_Preferred
Recombinant Protein;50UG;Human PCSK9, Mouse IgG2a Fc Tag (PC9-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # Q8NBP7-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 98.0 kDa.
Catalog No. 50-201-9543