Learn More
ACROBiosystemsSupplier Diversity Partner Human MUC-17 Protein, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner MU7H52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human MUC-17, His Tag (MU7-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 4131 - Leu 4390 (Accession # Q685J3-1).;This protein carries a polyhistidine tag at the C-terminus. This protein was cleaved within the SEA domain between 4243 Gly and 4244 Ser, and was cleaved into N and C-terminal fragment with calculated MW of 12.6 kDa and 18.6 kDa respectively. The protein migrates as 20 kDa and 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9553