Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) S protein RBD (G476S), His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SPDC52H4100UG
Recombinant Protein;100UG;SARS-CoV-2 S protein RBD (G476S), His Tag (SPD-C52H4) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1 (G476S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 32-35 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD (G476S), His Tag (Cat. No. SPD-C52H4) with a linear range of 2-16 ng/mL (QC tested).;Immobilized Anti-SARS-CoV-2 Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S protein RBD (G476S), His Tag (Cat. No. SPD-C52H4) with a linear range of 2-8 ng/mL (Routinely tested).;Human ACE2, Fc Tag (Cat. No. AC2-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind SARS-CoV-2 S protein RBD (G476S), His Tag (Cat. No. SPD-C52H4) with an affinity constant of
Catalog No. 50-201-9600