Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) S1 protein NTD, His Tag
Manufacturer:  ACROBiosystemsSupplier Diversity Partner S1DC52H6100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein NTD, His Tag (S1D-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized SARS-CoV-2 S1 protein NTD, His Tag (Cat. No. S1D-C52H6) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 S1 protein Antibody, human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-201-9602