Learn More
ACROBiosystems Biotinylated Human GUCY2C / Guanylyl cyclase C Protein, His,Avitag™
Supplier: ACROBiosystems GUCH82E925UG
Recombinant Protein;25UG;Biotinylated Human GUCY2C, His,Avitag (GUC-H82E9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Gln 430 (Accession # P25092-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 49.6 kDa. The protein migrates as 65-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human GUCY2C Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human GUCY2C, His,Avitag (Cat. No. GUC-H82E9) with a linear range of 0.2-6 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.