Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) Inhibitor screening Kit
Manufacturer:  ACROBiosystemsSupplier Diversity Partner EP10596TESTS
Encompass_Preferred
Kits;96TESTS;This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of SARS-CoV-2 inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized SARS-CoV-2 S protein RBD and in-house developed biotinylated Human ACE2 protein. This product is uniquely suitable for rapid high-throughput screening of SARS-CoV-2 inhibitors. Briefly, we provide you with a biotinylated Human ACE2 protein, a SARS-CoV-2 S protein RBD protein, an SARS-CoV-2 inhibitor (as method verified Reference), and Streptavidin-HRP reagent.
Catalog No. 50-201-9712