Learn More
ACROBiosystemsSupplier Diversity Partner ELISA Assay Kit for Anti-CTLA-4 h-mAb in Mouse Serum  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner ECMV196TESTS
Kits;96TESTS;The enclosed ELISA assay kit for anti-CTLA-4 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant CTLA-4 protein (ECD) and a biotinylated anti-CTLA-4 monoclonal antibody. The method employs the principle of competitive ELISA, allowing quantification of monoclonal anti-CTLA-4 antibodies in mouse serum.
Catalog No. 50-201-9830