Learn More
ACROBiosystems ELISA Assay Kit for Anti-HER-2 h-mAb in Mouse Serum
Supplier:  ACROBiosystems EHMV1480TESTS
Encompass_Preferred
Kits;480TESTS;The enclosed ELISA assay kit for anti-HER-2 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant HER-2 protein (ECD) and a biotinylated anti-HER-2 monoclonal antibody. The method employs the principle of competitive ELISA, allowing quantification of monoclonal anti-HER-2 antibodies in mouse serum.
Catalog No. 50-201-9836