Learn More
ACROBiosystemsSupplier Diversity Partner Human Erythropoietin R / EPO R Protein, Fc Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner EPRH5253100UG
Recombinant Protein;100UG;Human EPO R, Fc Tag (EPR-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Cys 241 (Accession # P19235-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.1 kDa.
Catalog No. 50-201-9870