Learn More
ACROBiosystems Human BTN1A1 / Butyrophilin Protein, Fc Tag
Supplier: ACROBiosystems BT1H5253100UG
Recombinant Protein;100UG;Human BTN1A1, Fc Tag (BT1-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # AAH96312).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.6 kDa. As a result of glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 116-130 kDa under non-reducing (NR) condition (SDS-PAGE).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.