Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) S protein RBD (K417N, E484K, N501Y), His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SPDC52HP100UG
Recombinant Protein;100UG;SARS-CoV-2 S protein RBD (K417N, E484K, N501Y), His Tag (SPD-C52Hp) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417N, E484K, N501Y)). K417N / E484K / N501Y mutations were identified in the SARS-CoV-2 variant (known as 20C/501Y.V2 or B.1.351 lineage) which emerged in South Africa.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.
Catalog No. 50-205-4737