Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein NTD (A222V), His Tag
Manufacturer:  ACROBiosystems S1DC52H8100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein NTD (A222V), His Tag (S1D-C52H8) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (A222V)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa.
Catalog No. 50-205-4747