Learn More
ACROBiosystemsSupplier Diversity Partner SARS-CoV-2 (COVID-19) S1 protein (Y144del), His Tag  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner S1NC52HC100UG
Recombinant Protein;100UG;SARS-CoV-2 S1 protein (Y144del), His Tag (S1N-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (Y144 del)). Y144 deletion was identified in the SARS-CoV-2 variant (known as 20B/501Y.V1, VOC 202012/01, or B.1.1.7 lineage) which emerged in the United Kindom.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.8 kDa.
Catalog No. 50-205-4788