Learn More
ACROBiosystems Human TREM2 Protein, Fc Tag
Supplier:  ACROBiosystems TR2H5254100UG
Encompass_Preferred
Recombinant Protein;100UG;Human TREM2, Fc Tag (TR2-H5254) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TREM2, Fc Tag (Cat. No. TR2-H5254) at 1 μg/mL (100 μL/well) can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-205-4828