Learn More
ACROBiosystems Biotinylated Human MARCO Protein, Avitag™,His Tag
Manufacturer:  ACROBiosystems MARH82E425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human MARCO, Avitag,His Tag (MAR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Val 65 - Val 520 (Accession # Q9UEW3-1).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 46.8 KDa.
Catalog No. 50-205-4850